This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03914999_s1
See other LOC102723919 GE Assays ›
Gene Symbol:
LOC102723919 LRIG2
Gene Name:
uncharacterized LOC102723919
leucine rich repeats and immunoglobulin like domains 2
Gene Aliases:
-
LIG-2, LIG2, UFS2
Chromosome Location:
Chr.1: 113131186 - 113132260 on Build GRCh38
Chromosome Location:
Chr.1: 113073170 - 113132260 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs03914999_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102723919
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK125443.1 - 2 1947 88
Gene Symbol: LRIG2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005271369.2 XP_005271426.1 17 10427 88
XM_017002952.1 XP_016858441.1 18 10399 88

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Downstream uncharacterized LOC643441 LOC643441 64.5kb Anti-sense Chr. 1 - 113196781 - 113206253 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

immunoglobulin superfamily cell adhesion molecule

Gene Ontology Categories:

Function(s)

sensory perception of sound
regulation of platelet-derived growth factor receptor signaling pathway
innervation

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000470000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000470000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000470000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000470000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000470000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000470000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0