This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03918240_s1
See other GTF2A1 GE Assays ›
Gene Symbol:
GTF2A1
Gene Name:
general transcription factor IIA subunit 1
Gene Aliases:
TF2A1, TFIIA, TFIIA-42, TFIIAL
Chromosome Location:
Chr.14: 81175452 - 81221231 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03918240_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GTF2A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001278940.1 NP_001265869.1 10 2206 105
NM_015859.3 NP_056943.1 9 2139 105
NM_201595.2 NP_963889.1 9 1759 105
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CD643192.1 - 1 351 Chr. 14 - 81179209 - 81179988 [-] on Build GRCh38 105
GenBankmRNA CN417534.1 - 1 155 Chr. 14 - 81179562 - 81179802 [-] on Build GRCh38 105
GenBankmRNA BX952230.1 - 1 122 Chr. 14 - 81179208 - 81179770 [-] on Build GRCh38 105
GenBankmRNA CN417537.1 - 1 331 Chr. 14 - 81179266 - 81179968 [-] on Build GRCh38 105
GenBankmRNA DA117173.1 - 1 71 Chr. 14 - 81179174 - 81179719 [-] on Build GRCh38 105
GenBankmRNA BC037828.1 - 1 355 Chr. 14 - 81176140 - 81180000 [-] on Build GRCh38 105
GenBankmRNA BM728666.1 - 1 337 Chr. 14 - 81179266 - 81179985 [-] on Build GRCh38 105
GenBankmRNA BM541549.1 - 1 340 Chr. 14 - 81178926 - 81179985 [-] on Build GRCh38 105
GenBankmRNA AI952296.1 - 1 Chr. 14 - 81179209 - 81179723 [+] on Build GRCh38 105

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream thyroid stimulating hormone receptor TSHR CHNG1,
LGR3,
hTSHR-I
-29.8kb Anti-sense Chr. 14 - 80954989 - 81146302 on NCBI Build GRCh38
Downstream small nucleolar RNA,
H/ACA box 79
SNORA79 ACA65,
SNORA79A
22.7kb Sense Chr. 14 - 81202695 - 81202834 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1400009834.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GTF2A1

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
snRNA transcription from RNA polymerase II promoter
DNA binding
transcription coactivator activity
protein binding
transcription factor binding
TBP-class protein binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline