This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03921576_s1
See other USP6 GE Assays ›
Gene Symbol:
USP6
Gene Name:
ubiquitin specific peptidase 6
Gene Aliases:
HRP1, TRE17, TRE2, TRESMCR, Tre-2, USP6-short
Chromosome Location:
Chr.17: 5116438 - 5175032 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03921576_s1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: USP6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017025290.1 XP_016880779.1 1 557 102
GenBank mRNA X78261.1 - 1050 102

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream zinc finger protein 232 ZNF232 ZSCAN11 -358.0 Anti-sense Chr. 17 - 5105734 - 5123116 on NCBI Build GRCh38
Downstream zinc finger protein 594 ZNF594 54.3kb Anti-sense Chr. 17 - 5174021 - 5191894 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017025290.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

cellular protein modification process
ubiquitin-dependent protein catabolic process
protein deubiquitination
regulation of vesicle-mediated transport
nucleic acid binding
cysteine-type endopeptidase activity
thiol-dependent ubiquitin-specific protease activity
protein binding
calmodulin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline