This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03921827_s1
See other FNIP2 GE Assays ›
Gene Symbol:
FNIP2
Gene Name:
folliculin interacting protein 2
Gene Aliases:
FNIPL, MAPO1
Chromosome Location:
Chr.4: 158768995 - 158908050 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03921827_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FNIP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005263160.3 XP_005263217.1 16 106
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA R95852.1 - Chr. - - [-] on Build 106
GenBankmRNA BX091857.1 - 1 254 Chr. 4 - 158872739 - 158873154 [-] on Build GRCh38 106
GenBankmRNA AF085902.1 - 1 254 Chr. 4 - 158872754 - 158873154 [-] on Build GRCh38 106
GenBankmRNA R97501.1 - Chr. - - [-] on Build 106

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream peptidylprolyl isomerase D PPID CYP-40,
CYPD
-149.4kb Sense Chr. 4 - 158709127 - 158723400 on NCBI Build GRCh38
Downstream chromosome 4 open reading frame 45 C4orf45 20.4kb Sense Chr. 4 - 158893532 - 159082883 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
regulation of protein phosphorylation
protein phosphorylation
intrinsic apoptotic signaling pathway in response to DNA damage
positive regulation of protein complex assembly
positive regulation of peptidyl-serine phosphorylation
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline