Assay Details
| Gene Symbol: CADM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC063484.1 | - | 1 | 1250 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 971 | LINC00971 | -89.4kb | Anti-sense | Chr. 3 - 84638405 - 84869575 on NCBI Build GRCh38 | |
| Downstream | microRNA 5688 | MIR5688 | 425.4kb | Sense | Chr. 3 - 85385710 - 85385792 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

CADM2
Genomic Map