Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC441369 | FLJ46284 | -89.2kb | Sense | Chr. 8 - 92712962 - 92786060 on NCBI Build GRCh38 | |
| Downstream | triple QxxK/R motif containing | TRIQK |
C8orf83, PRO0845, UPF0599 |
4.0kb | Sense | Chr. 8 - 92883530 - 92966150 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map