This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03989366_m1
See other IGSF22 GE Assays ›
Gene Symbol:
IGSF22
Gene Name:
immunoglobulin superfamily member 22
Gene Aliases:
IGFN2
Chromosome Location:
Chr.11: 18704305 - 18726939 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03989366_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGSF22
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_173588.3 NP_775859.3 18 - 19 3186 74
XM_011520030.2 XP_011518332.1 16 - 17 6090 74
XM_011520031.2 XP_011518333.1 15 - 16 5787 74
XM_017017607.1 XP_016873096.1 17 - 18 5981 74
XM_017017608.1 XP_016873097.1 17 - 18 4004 74
GenBank mRNA BC115396.1 - 18 - 19 3137 74
BC115397.1 - 17 - 18 3336 40068579 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100010276.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11001471.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IGSF22

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000412229.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000412229.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000412229.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000412229.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000412229.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000510673.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000510673.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11018620.hg.1 ENST00000513874.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11018620.hg.1 NM_173588.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11018620.hg.1 TC11001471.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1100178208.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100178208.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100178208.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay PSR1100178208.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100178208.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100178208.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay PSR1100178208.hg.1 TC1100010276.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000319338.6 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000412229.2 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000412229.2 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000412229.2 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000412229.2 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000412229.2 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000504981.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000510673.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000510673.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 ENST00000513874.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq
Affymetrix Human Clariom D Assay TC1100010276.hg.1 NM_173588.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline