This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04176261_m1
See other P2RX3 GE Assays ›
Gene Symbol:
P2RX3
Gene Name:
purinergic receptor P2X 3
Gene Aliases:
P2X3
Chromosome Location:
Chr.11: 57335943 - 57372399 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04176261_m1
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P2RX3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002559.3 NP_002550.2 1 263 107
XM_011545069.2 XP_011543371.1 2 924 107
XM_011545070.2 XP_011543372.1 2 230 107
XM_011545071.2 XP_011543373.1 1 798 107
GenBank mRNA AB016608.1 - 1 79 107
BC069162.1 - 1 155 7216977 107
BC074792.2 - 1 110 30915565 107
BC074793.2 - 1 110 30915348 107
BC109287.2 - 1 113 40031972 107
DB462670.1 - 1 263 107

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100007670.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11000461.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Homo sapiens 1700 Inventoried Amplicon spans exons and probe does not span exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene P2RX3

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
transport
signal transduction
neuromuscular synaptic transmission
blood coagulation
response to heat
response to cold
response to mechanical stimulus
response to carbohydrate
positive regulation of calcium ion transport into cytosol
urinary bladder smooth muscle contraction
neuronal action potential
peristalsis
response to ATP
purinergic nucleotide receptor signaling pathway
regulation of synaptic plasticity
positive regulation of calcium-mediated signaling
sensory perception of taste
protein homooligomerization
behavioral response to formalin induced pain
cation transmembrane transport
positive regulation of sensory perception of pain
purinergic nucleotide receptor activity
extracellular ATP-gated cation channel activity
ATP binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11005571.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11005571.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11005571.hg.1 TC11000461.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11005574.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11005574.hg.1 ENST00000439993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11005574.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11005574.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11005574.hg.1 TC11000461.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11005572.hg.1 ENST00000439993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11005572.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11005572.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11005572.hg.1 TC11000461.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay PSR1100153936.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100153936.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100153936.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100153936.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay PSR1100153936.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100153936.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100153936.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100153936.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay PSR1100153936.hg.1 TC1100007670.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay PSR1100153933.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100153933.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100153933.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay PSR1100153933.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100153933.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100153933.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay PSR1100153933.hg.1 TC1100007670.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100153935.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100153935.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100153935.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100153935.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay PSR1100153935.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100153935.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100153935.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100153935.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay PSR1100153935.hg.1 TC1100007670.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100153932.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100153932.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay PSR1100153932.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100153932.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay PSR1100153932.hg.1 TC1100007670.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 136497 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0