This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04176267_s1
See other ENPP2 GE Assays ›
Gene Symbol:
ENPP2
Gene Name:
ectonucleotide pyrophosphatase/phosphodiesterase 2
Gene Aliases:
ATX, ATX-X, AUTOTAXIN, LysoPLD, NPP2, PD-IALPHA, PDNP2
Chromosome Location:
Chr.8: 119557077 - 119673404 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04176267_s1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ENPP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001040092.2 NP_001035181.1 15 1376 108
NM_001130863.2 NP_001124335.1 15 1376 108
NM_006209.4 NP_006200.3 16 1532 108
NR_045555.1 4 293 108
XM_006716584.1 XP_006716647.1 16 1510 108
XM_006716585.1 XP_006716648.1 16 1510 108
XM_006716587.1 XP_006716650.1 16 1510 108
XM_017013571.1 XP_016869060.1 15 1410 108
XM_017013572.1 XP_016869061.1 15 1354 108
XM_017013573.1 XP_016869062.1 15 1410 108
XM_017013574.1 XP_016869063.1 15 1410 108
XM_017013575.1 XP_016869064.1 15 1354 108
GenBank mRNA AK130313.1 - 4 293 108
AK315253.1 - 15 1349 108
BC034961.2 - 15 1349 4828484 108
D45421.1 - 15 1323 108
DA919550.1 - 3 150 108
DA922156.1 - 4 328 108
EU131011.2 - 15 1290 108
L35594.1 - 16 1495 108

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ENPP2

Gene Ontology Categories:

Function(s) Process(es)

phosphate-containing compound metabolic process
receptor-mediated endocytosis
movement of cell or subcellular component
chemotaxis
immune response
G-protein coupled receptor signaling pathway
phospholipid catabolic process
positive regulation of epithelial cell migration
regulation of cell migration
phosphatidylcholine catabolic process
regulation of angiogenesis
positive regulation of peptidyl-tyrosine phosphorylation
nucleic acid phosphodiester bond hydrolysis
positive regulation of lamellipodium morphogenesis
nucleic acid binding
phosphodiesterase I activity
nucleotide diphosphatase activity
lysophospholipase activity
scavenger receptor activity
calcium ion binding
transcription factor binding
zinc ion binding
hydrolase activity
polysaccharide binding
alkylglycerophosphoethanolamine phosphodiesterase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline