This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04177464_m1
See other PLCB4 GE Assays ›
Gene Symbol:
PLCB4
Gene Name:
phospholipase C beta 4
Gene Aliases:
ARCND2, PI-PLC
Chromosome Location:
Chr.20: 9068710 - 9480816 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04177464_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLCB4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001172646.1 NP_001166117.1 2 - 3 211 90
NM_182797.2 NP_877949.2 1 - 2 58 90
XM_005260724.2 XP_005260781.1 2 - 3 252 90
XM_006723568.3 XP_006723631.1 3 - 4 234 90
XM_006723569.2 XP_006723632.1 2 - 3 332 90
XM_017027880.1 XP_016883369.1 2 - 3 252 90
XM_017027881.1 XP_016883370.1 3 - 4 234 90
XM_017027883.1 XP_016883372.1 2 - 3 252 90
GenBank mRNA AK122699.1 - 2 - 3 175 90
AK307482.1 - 2 - 3 120 90
AK307837.1 - 2 - 3 211 90
BI439140.1 - 3 - 4 257 90
DB270238.1 - 3 - 4 234 90

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001172646.1, NM_182797.2, XM_005260724.2, XM_006723568.3, XM_006723569.2, XM_017027880.1, XM_017027881.1, XM_017027883.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PLCB4

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

lipid catabolic process
intracellular signal transduction
negative regulation of potassium ion transport
inositol phosphate metabolic process
phosphatidylinositol phospholipase C activity
phospholipase C activity
signal transducer activity
calcium ion binding
protein binding
mitogen-activated protein kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0