This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04180561_g1
See other RALGAPA1 GE Assays ›
Gene Symbol:
RALGAPA1 RALGAPA1P1
Gene Name:
Ral GTPase activating protein catalytic alpha subunit 1
Ral GTPase activating protein catalytic alpha subunit 1 pseudogene 1
Gene Aliases:
GARNL1, GRIPE, RalGAPalpha1, TULIP1, p240
GARNL2, GARNL2P, RALGAPA1P, bA235C23.1
Chromosome Location:
Chr.14: 35538352 - 35809304 on Build GRCh38
Chromosome Location:
Chr.9: 105519824 - 105527319 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs04180561_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RALGAPA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001283043.1 NP_001269972.1 29 - 30 4400 94
NM_001283044.1 NP_001269973.1 29 - 30 4502 94
NM_014990.1 NP_055805.1 28 - 29 4361 94
NM_194301.2 NP_919277.2 28 - 29 4361 94
XM_005267491.3 XP_005267548.1 29 - 30 4580 94
XM_005267492.3 XP_005267549.1 29 - 30 4580 94
XM_005267493.3 XP_005267550.1 28 - 29 4439 94
XM_005267498.4 XP_005267555.1 14 - 15 2190 94
XM_006720098.2 XP_006720161.1 29 - 30 5957 94
XM_006720099.2 XP_006720162.1 29 - 30 5957 94
XM_006720100.3 XP_006720163.1 28 - 29 5816 94
XM_006720101.2 XP_006720164.1 29 - 30 5957 94
XM_006720102.3 XP_006720165.1 29 - 30 5663 94
XM_006720103.2 XP_006720166.1 29 - 30 4580 94
XM_011536615.2 XP_011534917.1 29 - 30 5957 94
XM_017021141.1 XP_016876630.1 28 - 29 5816 94
XM_017021142.1 XP_016876631.1 28 - 29 5816 94
XM_017021143.1 XP_016876632.1 29 - 30 5957 94
XM_017021144.1 XP_016876633.1 29 - 30 4580 94
XM_017021145.1 XP_016876634.1 28 - 29 4439 94
XM_017021146.1 XP_016876635.1 14 - 15 3626 94
XM_017021147.1 XP_016876636.1 14 - 15 2512 94
GenBank mRNA AB511280.1 - 29 - 30 4111 94
AL834362.1 - 5 - 6 472 94
AY596970.1 - 28 - 29 4361 94
AY596971.1 - 28 - 29 4361 94
BC042045.1 - 5 - 6 532 94
BC150596.1 - 29 - 30 4108 9056966 94
Gene Symbol: RALGAPA1P1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_104269.1 1 4400 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1400008945.hg.1
This assay targets the same location as HTA transcript cluster(s) TC14001038.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RALGAPA1
Click here to find siRNAs for Gene RALGAPA1P1

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
regulation of small GTPase mediated signal transduction
activation of GTPase activity
GTPase activator activity
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR14011875.hg.1 ENST00000258840.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14011875.hg.1 ENST00000307138.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14011875.hg.1 ENST00000335518.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14011875.hg.1 ENST00000382366.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14011875.hg.1 ENST00000389698.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14011875.hg.1 NM_014990.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14011875.hg.1 NM_194301.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14011875.hg.1 TC14001038.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1400165854.hg.1 ENST00000307138.10 Ensembl
Affymetrix Human Clariom D Assay PSR1400165854.hg.1 ENST00000382366.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400165854.hg.1 ENST00000389698.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400165854.hg.1 ENST00000553892.2 Ensembl
Affymetrix Human Clariom D Assay PSR1400165854.hg.1 NM_001283043.2 Refseq
Affymetrix Human Clariom D Assay PSR1400165854.hg.1 NM_001283044.2 Refseq
Affymetrix Human Clariom D Assay PSR1400165854.hg.1 NM_014990.2 Refseq
Affymetrix Human Clariom D Assay PSR1400165854.hg.1 NM_194301.3 Refseq
Affymetrix Human Clariom D Assay PSR1400165854.hg.1 ENST00000307138.10 Ensembl
Affymetrix Human Clariom D Assay PSR1400165854.hg.1 ENST00000382366.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400165854.hg.1 ENST00000389698.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400165854.hg.1 ENST00000553892.2 Ensembl
Affymetrix Human Clariom D Assay PSR1400165854.hg.1 NM_001283043.2 Refseq
Affymetrix Human Clariom D Assay PSR1400165854.hg.1 NM_001283044.2 Refseq
Affymetrix Human Clariom D Assay PSR1400165854.hg.1 NM_014990.2 Refseq
Affymetrix Human Clariom D Assay PSR1400165854.hg.1 NM_194301.3 Refseq
Affymetrix Human Clariom D Assay PSR1400165854.hg.1 TC1400008945.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0