This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04185351_m1
See other S100A7L2 GE Assays ›
Gene Symbol:
S100A7L2
Gene Name:
S100 calcium binding protein A7 like 2
Gene Aliases:
S100a7b
Chromosome Location:
Chr.1: 153436995 - 153440027 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04185351_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100A7L2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001045479.1 NP_001038944.2 2 - 3 249 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Process(es)

calcium ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline