This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04185561_m1
See other NAA60 GE Assays ›
Gene Symbol:
NAA60
Gene Name:
N(alpha)-acetyltransferase 60, NatF catalytic subunit
Gene Aliases:
HAT4, NAT15
Chromosome Location:
Chr.16: 3443611 - 3486963 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04185561_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAA60
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001083601.2 NP_001077070.1 2 - 3 301 69
GenBank mRNA AL833594.1 - 2 - 3 178 69
DC401343.1 - 2 - 3 301 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600011342.hg.1

Additional Information:

This assay is in the 5'UTR region of the NM_001083601.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

acetyltransferase

Gene Ontology Categories:

Function(s) Process(es)

nucleosome assembly
chromosome segregation
cell proliferation
N-terminal peptidyl-methionine acetylation
histone H4 acetylation
peptide alpha-N-acetyltransferase activity
H4 histone acetyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000360862.9 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000407558.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000414063.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000421765.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000424546.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000570372.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000570819.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000571107.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000572131.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000572169.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000572584.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000572739.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000572757.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000572942.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000573201.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000573580.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000573593.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000574328.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000574950.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000575076.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000575733.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000575936.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000576787.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000576916.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000577013.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 NM_001083600.2 Refseq
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 NM_001083601.2 Refseq
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 NM_024845.3 Refseq
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000360862.9 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000407558.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000414063.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000421765.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000424546.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000570372.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000570819.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000571107.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000572131.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000572169.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000572584.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000572739.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000572757.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000572942.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000573201.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000573580.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000573593.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000574328.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000574950.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000575076.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000575733.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000575936.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000576787.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000576916.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 ENST00000577013.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 NM_001083600.2 Refseq
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 NM_001083601.2 Refseq
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 NM_024845.3 Refseq
Affymetrix Human Clariom D Assay PSR1600189097.hg.1 TC1600011342.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0