This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04186202_s1
See other BDNF GE Assays ›
Gene Symbol:
BDNF
Gene Name:
brain derived neurotrophic factor
Gene Aliases:
ANON2, BULN2
Chromosome Location:
Chr.11: 27654893 - 27722058 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04186202_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BDNF
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_170735.5 NP_733931.1 1 1047 80
GenBank mRNA EF689009.1 - 1 1061 80
M61176.1 - 1 524 80

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the EF689009.1, M61176.1, NM_170735.5 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene BDNF

Panther Classification:

Molecular Function -

neurotrophic factor

Gene Ontology Categories:

Function(s) Process(es)

cell-cell signaling
nervous system development
axon guidance
synapse assembly
brain-derived neurotrophic factor receptor signaling pathway
positive regulation of brain-derived neurotrophic factor receptor signaling pathway
negative regulation of neuron apoptotic process
collateral sprouting
positive regulation of collateral sprouting
positive regulation of synapse assembly
neurotrophin TRKB receptor binding
growth factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline