This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04186514_m1
See other UBE2C GE Assays ›
Gene Symbol:
UBE2C
Gene Name:
ubiquitin conjugating enzyme E2 C
Gene Aliases:
UBCH10, dJ447F3.2
Chromosome Location:
Chr.20: 45812576 - 45816957 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04186514_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE2C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_104036.1 2 - 3 251 76
GenBank mRNA CB143441.1 - 2 - 3 169 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
negative regulation of cyclin-dependent protein serine/threonine kinase by cyclin degradation
exit from mitosis
free ubiquitin chain polymerization
protein ubiquitination
regulation of mitotic metaphase/anaphase transition
anaphase-promoting complex-dependent catabolic process
positive regulation of exit from mitosis
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
cell division
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
protein K48-linked ubiquitination
protein K11-linked ubiquitination
positive regulation of ubiquitin protein ligase activity
ubiquitin-protein transferase activity
protein binding
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0