This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04186565_m1
See other NDUFB6 GE Assays ›
Gene Symbol:
NDUFB6
Gene Name:
NADH:ubiquinone oxidoreductase subunit B6
Gene Aliases:
B17, CI
Chromosome Location:
Chr.9: 32553526 - 32573184 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04186565_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDUFB6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_182739.2 NP_877416.1 2 - 3 403 99
GenBank mRNA BI669356.1 - 2 - 3 382 99

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NDUFB6

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial electron transport, NADH to ubiquinone
mitochondrial respiratory chain complex I assembly
NADH dehydrogenase (ubiquinone) activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_182739.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0