This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04186724_m1
See other FAM173B GE Assays ›
Gene Symbol:
FAM173B
Gene Name:
family with sequence similarity 173 member B
Gene Aliases:
JS-2
Chromosome Location:
Chr.5: 10225507 - 10249909 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04186724_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FAM173B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001258388.1 NP_001245317.1 1 - 2 41 98
NM_001258389.1 NP_001245318.1 1 - 2 41 98
NM_199133.3 NP_954584.2 1 - 2 41 98
NR_047668.1 1 - 2 41 98
NR_047669.1 1 - 2 41 98
GenBank mRNA F29251.1 - 1 - 2 312 98

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001258388.1, NM_001258389.1, NM_199133.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene FAM173B

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline