Assay Details
| Gene Symbol: TNFRSF10D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003840.4 | NP_003831.2 | 3 - 4 | 470 | 73 | |
| GenBank mRNA | AF021232.1 | - | 3 - 4 | 453 | 73 | |
| AF021233.1 | - | 3 - 4 | 454 | 73 | ||
| AF023849.1 | - | 3 - 4 | 375 | 73 | ||
| AF029761.1 | - | 3 - 4 | 457 | 73 | ||
| AK313528.1 | - | 3 - 4 | 467 | 73 | ||
| AY358285.1 | - | 3 - 4 | 458 | 73 | ||
| BC052270.1 | - | 3 - 4 | 458 | 6572696 | 73 | |
| CD703247.1 | - | 3 - 4 | 502 | 73 | ||
More Information
Set Membership: |
|

TNFRSF10D
Genomic Map