This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04187869_g1
See other RAC1 GE Assays ›
Gene Symbol:
RAC1
Gene Name:
ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1)
Gene Aliases:
MIG5, Rac-1, TC-25, p21-Rac1
Chromosome Location:
Chr.7: 6374495 - 6403967 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04187869_g1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006908.4 NP_008839.2 3 - 4 461 73
GenBank mRNA AB451238.1 - 3 - 4 220 73
AB451363.1 - 3 - 4 220 73
AF498964.1 - 3 - 4 220 73
AK054993.1 - 3 - 4 436 73
AK291806.1 - 3 - 4 417 73
AY279384.1 - 3 - 4 378 73
BC004247.1 - 3 - 4 358 3610745 73
BC050687.1 - 3 - 4 408 6149377 73
BC107748.1 - 3 - 4 423 6699945 73
BQ689587.1 - 4 - 5 477 73
BT007121.1 - 3 - 4 220 73
M29870.1 - 3 - 4 220 73
M31467.1 - 3 - 4 220 73

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RAC1

Panther Classification:

Molecular Function -

small GTPase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
auditory receptor cell morphogenesis
mast cell chemotaxis
epithelial cell morphogenesis
movement of cell or subcellular component
inflammatory response
hyperosmotic response
cell adhesion
cell-matrix adhesion
G-protein coupled receptor signaling pathway
small GTPase mediated signal transduction
axon guidance
blood coagulation
cell proliferation
regulation of cell size
response to wounding
anatomical structure morphogenesis
regulation of hydrogen peroxide metabolic process
positive regulation of lamellipodium assembly
regulation of fibroblast migration
positive regulation of cell-substrate adhesion
regulation of neuron maturation
cerebral cortex radially oriented cell migration
embryonic olfactory bulb interneuron precursor migration
cerebral cortex GABAergic interneuron development
lamellipodium assembly
actin cytoskeleton organization
actin filament polymerization
platelet activation
regulation of cell migration
positive regulation of actin filament polymerization
T cell costimulation
ruffle organization
negative regulation of interleukin-23 production
substrate adhesion-dependent cell spreading
positive regulation of Rho protein signal transduction
intracellular signal transduction
Fc-epsilon receptor signaling pathway
Fc-gamma receptor signaling pathway involved in phagocytosis
positive regulation of apoptotic process
positive regulation of phosphatidylinositol 3-kinase activity
engulfment of apoptotic cell
cell-cell junction organization
bone resorption
positive regulation of DNA replication
vascular endothelial growth factor receptor signaling pathway
ephrin receptor signaling pathway
negative regulation of receptor-mediated endocytosis
anatomical structure arrangement
dendrite morphogenesis
cell motility
homeostasis of number of cells within a tissue
regulation of defense response to virus by virus
regulation of small GTPase mediated signal transduction
positive regulation of stress fiber assembly
localization within membrane
positive regulation of focal adhesion assembly
synaptic transmission, GABAergic
Wnt signaling pathway, planar cell polarity pathway
regulation of respiratory burst
cellular response to mechanical stimulus
semaphorin-plexin signaling pathway
dopaminergic neuron differentiation
protein localization to plasma membrane
positive regulation of neutrophil chemotaxis
cochlea morphogenesis
ruffle assembly
positive regulation of substrate adhesion-dependent cell spreading
GTPase activity
protein binding
GTP binding
Rab GTPase binding
enzyme binding
protein kinase binding
GTP-dependent protein binding
thioesterase binding
histone deacetylase binding
Rho GDP-dissociation inhibitor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000473564.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000473564.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000473564.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000473564.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000495499.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000495499.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000495499.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000495499.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000495499.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000473564.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000473564.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000473564.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000473564.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000495499.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000495499.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000495499.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000495499.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000495499.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0