This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04187890_g1
See other ATP5I GE Assays ›
Gene Symbol:
ATP5I
Gene Name:
ATP synthase, H+ transporting, mitochondrial Fo complex subunit E
Gene Aliases:
ATP5K
Chromosome Location:
Chr.4: 672436 - 674338 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04187890_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP5I
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007100.3 NP_009031.1 2 - 3 182 90
GenBank mRNA AB028624.1 - 2 - 3 134 90
AK000656.1 - 2 - 3 97 90
BC003679.1 - 2 - 3 118 4052832 90
BC105610.1 - 2 - 3 124 6739801 90
BG940181.1 - 2 - 3 110 90
BT006922.1 - 2 - 3 91 90
D50371.1 - 2 - 3 154 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400009694.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04000951.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ATP5I

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

ATP biosynthetic process
mitochondrial ATP synthesis coupled proton transport
hydrogen ion transmembrane transporter activity
ATPase activity
transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR04012783.hg.1 TC04000951.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04012783.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04012783.hg.1 ENST00000506525.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04012783.hg.1 NM_007100.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04012783.hg.1 NR_033743.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04012789.hg.1 TC04000951.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04012789.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04012789.hg.1 NM_007100.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04012789.hg.1 NR_033743.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04012791.hg.1 TC04000951.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04012791.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04012791.hg.1 NM_007100.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04012791.hg.1 NR_033743.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 ENST00000505852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 ENST00000505852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 ENST00000505852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 ENST00000505852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 ENST00000505852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 ENST00000506525.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 ENST00000506525.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 ENST00000506525.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 ENST00000515116.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 ENST00000515116.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 ENST00000515202.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 ENST00000515202.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 ENST00000515202.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 ENST00000515202.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 NM_007100.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 NM_007100.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 NM_007100.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 NM_007100.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 NM_007100.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 NM_007100.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 NR_033743.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 NR_033743.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 NR_033743.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 NR_033743.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009694.hg.1 NR_033743.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04012787.hg.1 TC04000951.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04012787.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04012787.hg.1 NM_007100.3 Refseq
Affymetrix Human Clariom D Assay PSR0400166297.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400166297.hg.1 ENST00000505852.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400166297.hg.1 NM_007100.3 Refseq
Affymetrix Human Clariom D Assay PSR0400166297.hg.1 NR_033743.1 Refseq
Affymetrix Human Clariom D Assay PSR0400166297.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400166297.hg.1 ENST00000505852.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400166297.hg.1 NM_007100.3 Refseq
Affymetrix Human Clariom D Assay PSR0400166297.hg.1 NR_033743.1 Refseq
Affymetrix Human Clariom D Assay PSR0400166297.hg.1 TC0400009694.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0400166293.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400166293.hg.1 ENST00000515202.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400166293.hg.1 NM_007100.3 Refseq
Affymetrix Human Clariom D Assay PSR0400166293.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400166293.hg.1 ENST00000515202.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400166293.hg.1 NM_007100.3 Refseq
Affymetrix Human Clariom D Assay PSR0400166293.hg.1 TC0400009694.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0400166296.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400166296.hg.1 ENST00000505852.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400166296.hg.1 NM_007100.3 Refseq
Affymetrix Human Clariom D Assay PSR0400166296.hg.1 NR_033743.1 Refseq
Affymetrix Human Clariom D Assay PSR0400166296.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400166296.hg.1 ENST00000505852.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400166296.hg.1 NM_007100.3 Refseq
Affymetrix Human Clariom D Assay PSR0400166296.hg.1 NR_033743.1 Refseq
Affymetrix Human Clariom D Assay PSR0400166296.hg.1 TC0400009694.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0400009694.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Clariom D Assay TC0400009694.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Clariom D Assay TC0400009694.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Clariom D Assay TC0400009694.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Clariom D Assay TC0400009694.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Clariom D Assay TC0400009694.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Clariom D Assay TC0400009694.hg.1 ENST00000505852.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009694.hg.1 ENST00000505852.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009694.hg.1 ENST00000505852.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009694.hg.1 ENST00000505852.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009694.hg.1 ENST00000505852.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009694.hg.1 ENST00000506525.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009694.hg.1 ENST00000506525.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009694.hg.1 ENST00000506525.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009694.hg.1 ENST00000515116.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009694.hg.1 ENST00000515116.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009694.hg.1 ENST00000515202.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009694.hg.1 ENST00000515202.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009694.hg.1 ENST00000515202.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009694.hg.1 ENST00000515202.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009694.hg.1 NM_007100.3 Refseq
Affymetrix Human Clariom D Assay TC0400009694.hg.1 NM_007100.3 Refseq
Affymetrix Human Clariom D Assay TC0400009694.hg.1 NM_007100.3 Refseq
Affymetrix Human Clariom D Assay TC0400009694.hg.1 NM_007100.3 Refseq
Affymetrix Human Clariom D Assay TC0400009694.hg.1 NM_007100.3 Refseq
Affymetrix Human Clariom D Assay TC0400009694.hg.1 NM_007100.3 Refseq
Affymetrix Human Clariom D Assay TC0400009694.hg.1 NR_033743.1 Refseq
Affymetrix Human Clariom D Assay TC0400009694.hg.1 NR_033743.1 Refseq
Affymetrix Human Clariom D Assay TC0400009694.hg.1 NR_033743.1 Refseq
Affymetrix Human Clariom D Assay TC0400009694.hg.1 NR_033743.1 Refseq
Affymetrix Human Clariom D Assay TC0400009694.hg.1 NR_033743.1 Refseq
Affymetrix Human Clariom D Assay PSR0400166288.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400166288.hg.1 ENST00000505852.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400166288.hg.1 ENST00000506525.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400166288.hg.1 ENST00000515116.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400166288.hg.1 ENST00000515202.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400166288.hg.1 NM_007100.3 Refseq
Affymetrix Human Clariom D Assay PSR0400166288.hg.1 NR_033743.1 Refseq
Affymetrix Human Clariom D Assay PSR0400166288.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400166288.hg.1 ENST00000505852.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400166288.hg.1 ENST00000506525.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400166288.hg.1 ENST00000515116.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400166288.hg.1 ENST00000515202.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400166288.hg.1 NM_007100.3 Refseq
Affymetrix Human Clariom D Assay PSR0400166288.hg.1 NR_033743.1 Refseq
Affymetrix Human Clariom D Assay PSR0400166288.hg.1 TC0400009694.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0400166299.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400166299.hg.1 ENST00000505852.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400166299.hg.1 NM_007100.3 Refseq
Affymetrix Human Clariom D Assay PSR0400166299.hg.1 NR_033743.1 Refseq
Affymetrix Human Clariom D Assay PSR0400166299.hg.1 ENST00000304312.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400166299.hg.1 ENST00000505852.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400166299.hg.1 NM_007100.3 Refseq
Affymetrix Human Clariom D Assay PSR0400166299.hg.1 NR_033743.1 Refseq
Affymetrix Human Clariom D Assay PSR0400166299.hg.1 TC0400009694.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0