This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04187949_m1
See other NAT6 GE Assays ›
Gene Symbol:
NAT6
Gene Name:
N-acetyltransferase 6
Gene Aliases:
FUS-2, FUS2
Chromosome Location:
Chr.3: 50296402 - 50299421 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04187949_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAT6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012191.3 NP_036323.2 1 - 2 212 67
GenBank mRNA BC004483.2 - 1 - 2 72 3689733 67

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC004483.2, NM_012191.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NAT6

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Process(es)

N-acetyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0