This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04188017_g1
See other NENF GE Assays ›
Gene Symbol:
NENF
Gene Name:
neudesin neurotrophic factor
Gene Aliases:
CIR2, SCIRP10, SPUF
Chromosome Location:
Chr.1: 212432887 - 212446379 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04188017_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NENF
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013349.4 NP_037481.1 2 - 3 292 96
GenBank mRNA AB126219.1 - 2 - 3 235 96
AF049672.1 - 2 - 3 90 96
AF173937.1 - 2 - 3 295 96
AK074431.1 - 2 - 3 260 96
AK223135.1 - 2 - 3 260 96
AY762102.1 - 2 - 3 251 96
BC008823.2 - 2 - 3 241 4303614 96

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NENF

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of appetite
positive regulation of MAPK cascade
growth factor activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0