This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04188104_g1
See other CCT8L2 GE Assays ›
Gene Symbol:
CCT8L2
Gene Name:
chaperonin containing TCP1 subunit 8 like 2
Gene Aliases:
CESK1
Chromosome Location:
Chr.22: 16590758 - 16592810 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04188104_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CCT8L2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014406.4 NP_055221.1 1 149 89
GenBank mRNA BC033797.1 - 1 149 5172617 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2200007871.hg.1
This assay targets the same location as HTA transcript cluster(s) TC22000461.hg.1

Additional Information:

This assay is in the 5'UTR region of the BC033797.1,NM_014406.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CCT8L2

Panther Classification:

Molecular Function -

chaperonin

Gene Ontology Categories:

Function(s) Process(es)

transport
potassium ion transmembrane transport
anion transmembrane transport
anion channel activity
ATP binding
calcium-activated potassium channel activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR22008341.hg.1 ENST00000359963.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22008341.hg.1 NM_014406.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22008341.hg.1 TC22000461.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR22008340.hg.1 ENST00000359963.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22008340.hg.1 NM_014406.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22008340.hg.1 TC22000461.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2200007871.hg.1 ENST00000359963.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007871.hg.1 ENST00000359963.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007871.hg.1 ENST00000359963.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007871.hg.1 ENST00000359963.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007871.hg.1 ENST00000359963.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007871.hg.1 NM_014406.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007871.hg.1 NM_014406.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007871.hg.1 NM_014406.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007871.hg.1 NM_014406.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007871.hg.1 NM_014406.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22008342.hg.1 ENST00000359963.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22008342.hg.1 NM_014406.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22008342.hg.1 TC22000461.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR2200155903.hg.1 TC2200007871.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2200155903.hg.1 ENST00000359963.4 Ensembl
Affymetrix Human Clariom D Assay PSR2200155903.hg.1 NM_014406.4 Refseq
Affymetrix Human Clariom D Assay PSR2200155903.hg.1 ENST00000359963.4 Ensembl
Affymetrix Human Clariom D Assay PSR2200155903.hg.1 NM_014406.4 Refseq
Affymetrix Human Clariom D Assay PSR2200155902.hg.1 TC2200007871.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2200155902.hg.1 ENST00000359963.4 Ensembl
Affymetrix Human Clariom D Assay PSR2200155902.hg.1 NM_014406.4 Refseq
Affymetrix Human Clariom D Assay PSR2200155902.hg.1 ENST00000359963.4 Ensembl
Affymetrix Human Clariom D Assay PSR2200155902.hg.1 NM_014406.4 Refseq
Affymetrix Human Clariom D Assay PSR2200155900.hg.1 TC2200007871.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2200155900.hg.1 ENST00000359963.4 Ensembl
Affymetrix Human Clariom D Assay PSR2200155900.hg.1 NM_014406.4 Refseq
Affymetrix Human Clariom D Assay PSR2200155900.hg.1 ENST00000359963.4 Ensembl
Affymetrix Human Clariom D Assay PSR2200155900.hg.1 NM_014406.4 Refseq
Affymetrix Human Clariom D Assay TC2200007871.hg.1 ENST00000359963.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007871.hg.1 ENST00000359963.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007871.hg.1 ENST00000359963.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007871.hg.1 ENST00000359963.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007871.hg.1 ENST00000359963.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007871.hg.1 NM_014406.4 Refseq
Affymetrix Human Clariom D Assay TC2200007871.hg.1 NM_014406.4 Refseq
Affymetrix Human Clariom D Assay TC2200007871.hg.1 NM_014406.4 Refseq
Affymetrix Human Clariom D Assay TC2200007871.hg.1 NM_014406.4 Refseq
Affymetrix Human Clariom D Assay TC2200007871.hg.1 NM_014406.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0