This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04189541_g1
See other ECE2 GE Assays ›
Gene Symbol:
ECE2
Gene Name:
endothelin converting enzyme 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 184249657 - 184293031 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04189541_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ECE2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_032331.3 NP_115707.2 2 - 3 523 95
GenBank mRNA BC005835.2 - 2 - 3 519 95
BC012449.1 - 2 - 3 496 95
BC069005.1 - 2 - 3 509 95
BU568980.1 - 526 95

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ECE2

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

cell-cell signaling
brain development
heart development
cardioblast differentiation
peptide hormone processing
methylation
metalloendopeptidase activity
protein binding
methyltransferase activity
metal ion binding

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 193716 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0