This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04189783_m1
See other CD200R1 GE Assays ›
Gene Symbol:
CD200R1
Gene Name:
CD200 receptor 1
Gene Aliases:
CD200R, HCRTR2, MOX2R, OX2R
Chromosome Location:
Chr.3: 112922685 - 112975090 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04189783_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CD200R1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138806.3 NP_620161.1 1 - 2 302 83
NM_138939.2 NP_620385.1 1 - 2 302 83
GenBank mRNA AF495380.1 - 1 - 2 124 83
AF497549.1 - 1 - 2 124 83
AK293071.1 - 1 - 2 247 83
AY284975.1 - 1 - 2 291 83
AY358910.1 - 1 - 2 327 83
BC069661.1 - 1 - 2 100 7262527 83
BC069721.1 - 1 - 2 100 7262539 83
BC069743.1 - 1 - 2 100 7262551 83
BC143393.1 - 1 - 2 148 9051899 83

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CD200R1

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

viral process
regulation of immune response
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline