| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03029821.hg.1 |
ENST00000308611.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03029821.hg.1 |
ENST00000440122.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03029821.hg.1 |
ENST00000471858.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03029821.hg.1 |
ENST00000490004.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03029821.hg.1 |
NM_138806.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03029821.hg.1 |
NM_138939.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03029821.hg.1 |
NM_138940.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03029821.hg.1 |
NM_170780.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03029821.hg.1 |
TC03001654.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300189403.hg.1 |
ENST00000308611.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300189403.hg.1 |
ENST00000440122.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300189403.hg.1 |
ENST00000471858.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300189403.hg.1 |
ENST00000490004.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300189403.hg.1 |
NM_138806.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300189403.hg.1 |
NM_138939.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300189403.hg.1 |
NM_138940.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300189403.hg.1 |
NM_170780.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300189403.hg.1 |
ENST00000308611.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300189403.hg.1 |
ENST00000440122.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300189403.hg.1 |
ENST00000471858.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300189403.hg.1 |
ENST00000490004.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300189403.hg.1 |
NM_138806.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300189403.hg.1 |
NM_138939.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300189403.hg.1 |
NM_138940.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300189403.hg.1 |
NM_170780.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300189403.hg.1 |
TC0300012007.hg.1 |
Clariom D TC |