This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04191876_gH
See other TMEM183A GE Assays ›
Gene Symbol:
TMEM183A TMEM183B
Gene Name:
transmembrane protein 183A
transmembrane protein 183B
Gene Aliases:
C1orf37
C1orf37-DUP
Chromosome Location:
Chr.1: 203007367 - 203024848 on Build GRCh38
Chromosome Location:
Chr.3: 149981661 - 149983366 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs04191876_gH
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEM183A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001322955.1 NP_001309884.1 5 - 6 799 89
NM_001322956.1 NP_001309885.1 5 - 6 799 89
NM_001322957.1 NP_001309886.1 5 - 6 802 89
NM_001322958.1 NP_001309887.1 4 - 5 712 89
NM_001322959.1 NP_001309888.1 4 - 5 709 89
NM_138391.4 NP_612400.3 5 - 6 763 89
GenBank mRNA AF070537.1 - 5 - 6 780 89
AK291426.1 - 5 - 6 777 89
AK312985.1 - 5 - 6 777 89
BC013073.1 - 1 763 4151716 89
BC038952.2 - 1 815 5740484 89
BC150647.1 - 5 - 6 745 9057018 89
Gene Symbol: TMEM183B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001079809.1 NP_001073277.1 1 761 89
GenBank mRNA AK297222.1 - 5 - 6 681 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100011242.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01001681.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TMEM183A
Click here to find siRNAs for Gene TMEM183B

Gene Ontology Categories:

Process(es)

protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01026079.hg.1 TC01001681.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01026079.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01026079.hg.1 NM_001079809.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01026079.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01026076.hg.1 TC01001681.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01026076.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01026076.hg.1 ENST00000468449.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01026076.hg.1 NM_001079809.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01026076.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000463015.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000463015.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000463015.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000488097.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000488097.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000488097.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000488097.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000488097.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay PSR0100182504.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100182504.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay PSR0100182504.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100182504.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay PSR0100182504.hg.1 TC0100011242.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100182502.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100182502.hg.1 ENST00000488097.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100182502.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay PSR0100182502.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100182502.hg.1 ENST00000488097.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100182502.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay PSR0100182502.hg.1 TC0100011242.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000463015.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000463015.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000463015.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000488097.2 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000488097.2 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000488097.2 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000488097.2 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000488097.2 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0