Assay Details
| Gene Symbol: DEFB103B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018661.4 | NP_061131.1 | 2 | 434 | 88 | |
| GenBank mRNA | AB037972.1 | - | 2 | 239 | 88 | |
| AJ237673.1 | - | 2 | 248 | 88 | ||
| Gene Symbol: DEFB103A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081551.3 | NP_001075020.1 | 2 | 434 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 566828 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 301920 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P169017 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P169017 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 4495 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 6431 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 36847 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P169017 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P169017 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P169017 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P169017 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8149173 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8149176 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8144477 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8149174 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8144479 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8149175 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8144476 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8144480 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8144478 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8149177 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8149172 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8144473 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006623.hg.1 | ENST00000314357.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006623.hg.1 | ENST00000314357.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006623.hg.1 | ENST00000314357.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006623.hg.1 | ENST00000314357.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009464.hg.1 | ENST00000318124.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009464.hg.1 | ENST00000318124.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009464.hg.1 | ENST00000318124.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009464.hg.1 | ENST00000318124.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009464.hg.1 | NM_001081551.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009464.hg.1 | NM_001081551.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009464.hg.1 | NM_001081551.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009464.hg.1 | NM_001081551.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009464.hg.1 | NM_018661.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009464.hg.1 | NM_018661.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009464.hg.1 | NM_018661.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009464.hg.1 | NM_018661.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800164721.hg.1 | ENST00000318124.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800164721.hg.1 | NM_001081551.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800164721.hg.1 | NM_018661.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800164721.hg.1 | ENST00000318124.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800164721.hg.1 | NM_001081551.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800164721.hg.1 | NM_018661.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800164721.hg.1 | TC0800009464.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800006623.hg.1 | ENST00000314357.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006623.hg.1 | ENST00000314357.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006623.hg.1 | ENST00000314357.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006623.hg.1 | ENST00000314357.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009464.hg.1 | ENST00000318124.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009464.hg.1 | ENST00000318124.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009464.hg.1 | ENST00000318124.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009464.hg.1 | ENST00000318124.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009464.hg.1 | NM_001081551.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009464.hg.1 | NM_001081551.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009464.hg.1 | NM_001081551.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009464.hg.1 | NM_001081551.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009464.hg.1 | NM_018661.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009464.hg.1 | NM_018661.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009464.hg.1 | NM_018661.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009464.hg.1 | NM_018661.4 | Refseq |
| Human Genome U133 Plus 2.0 Array | 224239_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 224239_at | - | - |
| Human Genome U133 Set | 224239_PM_at | - | - |
| Human Genome U133 Set | 224239_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035490 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035490 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 100289 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_2132515 | NM_018661.3 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2132515 | AB037972.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2132515 | AJ237673.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1684308 | NM_001081551.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1684308 | AF295370.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1684308 | AF301470.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1684308 | NM_018661.3 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1684308 | AB037972.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2132515 | NM_018661.3 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2132515 | AB037972.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2132515 | AJ237673.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2132515 | NM_018661.3 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2132515 | AB037972.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2132515 | AJ237673.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2132515 | NM_018661.3 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2132515 | AB037972.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2132515 | AJ237673.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_13124886-S | NM_001081551.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_13124886-S | AF295370.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_13124886-S | AF301470.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_13124886-S | NM_018661.3 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_13124886-S | AB037972.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_20941-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0002370292 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0002370292 | - | - |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 270017 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_20941 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2132515 | NM_018661.3 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2132515 | AB037972.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2132515 | AJ237673.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2132515 | NM_018661.3 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2132515 | AB037972.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2132515 | AJ237673.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1684308 | NM_001081551.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1684308 | AF295370.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1684308 | AF301470.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1684308 | NM_018661.3 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1684308 | AB037972.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2132515 | NM_018661.3 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2132515 | AB037972.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2132515 | AJ237673.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2132515 | NM_018661.3 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2132515 | AB037972.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2132515 | AJ237673.1 | human_transcriptome |

DEFB103B
Genomic Map