This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04231571_s1
See other MIR339 GE Assays ›
Gene Symbol:
MIR339
Gene Name:
microRNA 339
Gene Aliases:
MIRN339, hsa-mir-339, mir-339
Chromosome Location:
Chr.7: 1022933 - 1023026 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04231571_s1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer:
434.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR339
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029898.1 1 52 69
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA N27306.1 - 1 48 Chr. 7 - 1022517 - 1023022 [-] on Build GRCh38 69
GenBankmRNA AI421412.1 - 1 48 Chr. 7 - 1022543 - 1023022 [-] on Build GRCh38 69

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream chromosome 7 open reading frame 50 C7orf50 YCR016W Sense Chr. 7 - 978485 - 1138291 on NCBI Build GRCh38
Downstream G protein-coupled receptor 146 GPR146 PGR8 21.5kb Anti-sense Chr. 7 - 1044573 - 1059269 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC0700009998.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07001078.hg.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25472983 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261046 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25472999 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25473011 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261060 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261047 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261037 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261016 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261028 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261033 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15898656 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15898626 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261051 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25472967 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15898652 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261024 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15898662 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261038 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261050 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261056 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261040 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261054 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261021 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261017 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261044 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15898654 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25472979 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261049 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261019 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261014 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261036 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261027 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15898650 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15898655 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261018 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261020 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25472975 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15898658 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261048 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15898658 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261060 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25472975 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261014 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261028 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261050 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261024 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25472983 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261038 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15898662 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261048 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261036 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25473011 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261017 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261033 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15898626 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261027 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261046 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15898652 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15898656 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261047 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261040 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261016 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15898654 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25472979 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25472999 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261049 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261021 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261037 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261018 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15898650 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261051 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261044 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261054 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25472967 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261019 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261020 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15898655 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05261056 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.1 ST Array 8137707 - -
Affymetrix Human Transcriptome Array 2.0 PSR07016572.hg.1 ENST00000362153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07016572.hg.1 NR_029898.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07016572.hg.1 TC07001078.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0700009998.hg.1 ENST00000362153.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009998.hg.1 NR_029898.1 Refseq
Affymetrix Human Clariom D Assay TC0700009998.hg.1 ENST00000362153.2 Ensembl
Affymetrix Human Clariom D Assay TC0700009998.hg.1 NR_029898.1 Refseq
Affymetrix Human Clariom D Assay PSR0700171152.hg.1 ENST00000362153.2 Ensembl
Affymetrix Human Clariom D Assay PSR0700171152.hg.1 NR_029898.1 Refseq
Affymetrix Human Clariom D Assay PSR0700171152.hg.1 ENST00000362153.2 Ensembl
Affymetrix Human Clariom D Assay PSR0700171152.hg.1 NR_029898.1 Refseq
Affymetrix Human Clariom D Assay PSR0700171152.hg.1 TC0700009998.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3309189 NR_029898.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3309189 NR_029898.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline