This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04231808_s1
See other MIR3165 GE Assays ›
Gene Symbol:
MIR3165
Gene Name:
microRNA 3165
Gene Aliases:
mir-3165
Chromosome Location:
Chr.11: 72072228 - 72072302 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04231808_s1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer:
434.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR3165
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_036123.1 1 33 70

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC100128494 LOC100128494 -51.3kb Anti-sense Chr. 11 - 72014291 - 72020910 on NCBI Build GRCh38
Downstream leucine rich transmembrane and O-methyltransferase domain containing LRTOMT CFAP111,
DFNB63,
LRRC51
8.0kb Anti-sense Chr. 11 - 72079982 - 72110782 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC1100011541.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11002051.hg.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11024483.hg.1 NR_036123.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11024483.hg.1 TC11002051.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1100011541.hg.1 ENST00000579874.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011541.hg.1 NR_036123.1 Refseq
Affymetrix Human Clariom D Assay PSR1100188858.hg.1 ENST00000579874.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100188858.hg.1 NR_036123.1 Refseq
Affymetrix Human Clariom D Assay PSR1100188858.hg.1 ENST00000579874.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100188858.hg.1 NR_036123.1 Refseq
Affymetrix Human Clariom D Assay PSR1100188858.hg.1 TC1100011541.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100011541.hg.1 ENST00000579874.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011541.hg.1 NR_036123.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline