This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04231940_g1
See other VPS9D1-AS1 GE Assays ›
Gene Symbol:
VPS9D1-AS1
Gene Name:
VPS9D1 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.16: 89711856 - 89718165 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04231940_g1
Size
Availability Made To Order
Catalog # 4426961
Price 416.00
Your Price
Online offer: 390.65
390.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VPS9D1-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_036480.1 1 - 2 366 80
GenBank mRNA AK096286.1 - 1 - 2 367 80
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BI086820.1 - 1 - 2 349 Chr. 16 - 89711873 - 89716638 [+] on Build GRCh38 80
GenBankmRNA BM909691.1 - 140 Chr. - - [-] on Build 80
GenBankmRNA HY258205.1 - 1 - 2 288 Chr. 16 - 89711972 - 89716565 [+] on Build GRCh38 80
GenBankmRNA DA738595.1 - 1 - 2 366 Chr. 16 - 89711856 - 89716677 [+] on Build GRCh38 80
GenBankmRNA BF315011.1 - 1 - 2 334 Chr. 16 - 89711890 - 89716755 [+] on Build GRCh38 80
GenBankmRNA BE312532.1 - 1 - 2 315 Chr. 16 - 89711908 - 89716625 [+] on Build GRCh38 80
GenBankmRNA DA699958.1 - 1 - 2 367 Chr. 16 - 89711856 - 89716495 [+] on Build GRCh38 80

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream spermatogenesis associated 2 like SPATA2L C16orf76,
tamo
-10.1kb Anti-sense Chr. 16 - 89696357 - 89701993 on NCBI Build GRCh38
Downstream zinc finger protein 276 ZNF276 CENP-Z,
CENPZ,
ZADT,
ZFP276,
ZNF477
7.4kb Sense Chr. 16 - 89720368 - 89740925 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600008898.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000704.hg.1

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16010005.hg.1 NR_036480.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16010005.hg.1 TC16000704.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR16010006.hg.1 NR_036480.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16010006.hg.1 TC16000704.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1600008898.hg.1 ENST00000562298.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008898.hg.1 ENST00000562298.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008898.hg.1 ENST00000562866.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008898.hg.1 ENST00000562866.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008898.hg.1 ENST00000562866.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008898.hg.1 ENST00000562866.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008898.hg.1 ENST00000562866.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008898.hg.1 ENST00000562866.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008898.hg.1 NR_036480.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008898.hg.1 NR_036480.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008898.hg.1 NR_036480.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008898.hg.1 NR_036480.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008898.hg.1 NR_036480.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008898.hg.1 NR_036480.1 Refseq
Affymetrix Human Clariom D Assay PSR1600166603.hg.1 ENST00000562866.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600166603.hg.1 NR_036480.1 Refseq
Affymetrix Human Clariom D Assay PSR1600166603.hg.1 ENST00000562866.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600166603.hg.1 NR_036480.1 Refseq
Affymetrix Human Clariom D Assay PSR1600166603.hg.1 TC1600008898.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600166604.hg.1 ENST00000562866.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600166604.hg.1 NR_036480.1 Refseq
Affymetrix Human Clariom D Assay PSR1600166604.hg.1 ENST00000562866.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600166604.hg.1 NR_036480.1 Refseq
Affymetrix Human Clariom D Assay PSR1600166604.hg.1 TC1600008898.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1600008898.hg.1 ENST00000562298.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008898.hg.1 ENST00000562298.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008898.hg.1 ENST00000562866.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008898.hg.1 ENST00000562866.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008898.hg.1 ENST00000562866.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008898.hg.1 ENST00000562866.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008898.hg.1 ENST00000562866.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008898.hg.1 ENST00000562866.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008898.hg.1 NR_036480.1 Refseq
Affymetrix Human Clariom D Assay TC1600008898.hg.1 NR_036480.1 Refseq
Affymetrix Human Clariom D Assay TC1600008898.hg.1 NR_036480.1 Refseq
Affymetrix Human Clariom D Assay TC1600008898.hg.1 NR_036480.1 Refseq
Affymetrix Human Clariom D Assay TC1600008898.hg.1 NR_036480.1 Refseq
Affymetrix Human Clariom D Assay TC1600008898.hg.1 NR_036480.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mlg7h:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline