This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04232067_m1
See other GPR33 GE Assays ›
Gene Symbol:
GPR33
Gene Name:
G protein-coupled receptor 33 (gene/pseudogene)
Gene Aliases:
-
Chromosome Location:
Chr.14: 31482944 - 31487945 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04232067_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR33
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001197184.2 NP_001184113.2 1 - 2 47 82
NR_036675.2 1 - 2 47 82
GenBank mRNA GQ981321.1 - 1 - 2 47 82
GQ981322.1 - 1 - 2 47 82

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the GQ981322.1, NM_001197184.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor Homo sapiens Inventoried Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1400008857.hg.1 ENST00000399285.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008857.hg.1 ENST00000399285.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008857.hg.1 NM_001197184.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008857.hg.1 NM_001197184.2 Refseq
Affymetrix Human Clariom D Assay TC1400008857.hg.1 ENST00000399285.3 Ensembl
Affymetrix Human Clariom D Assay TC1400008857.hg.1 ENST00000399285.3 Ensembl
Affymetrix Human Clariom D Assay TC1400008857.hg.1 NM_001197184.2 Refseq
Affymetrix Human Clariom D Assay TC1400008857.hg.1 NM_001197184.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline