This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04233524_s1
See other MIR107 GE Assays ›
Gene Symbol:
MIR107
Gene Name:
microRNA 107
Gene Aliases:
MIRN107, miR-107
Chromosome Location:
Chr.10: 89592747 - 89592827 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04233524_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR107
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029524.1 1 41 77
GenBank mRNA AF480550.1 - 1 41 77

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream solute carrier family 16 member 12 SLC16A12 CJMG,
CRT2,
MCT12
-57.2kb Sense Chr. 10 - 89430294 - 89536028 on NCBI Build GRCh38
Downstream tigger transposable element derived 2 pseudogene FLJ37201 98.5kb Sense Chr. 10 - 89691300 - 89697928 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC1000011383.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001508.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773291 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773300 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773304 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384090 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384095 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384086 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773337 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773297 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384045 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773306 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773319 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16371377 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384097 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384096 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773311 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773294 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773326 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384073 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773317 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773287 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384098 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773292 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773318 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773299 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773322 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384081 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773325 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773340 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773315 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773296 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384094 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16371381 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773316 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773303 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773285 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384103 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773309 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16371378 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16371375 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773320 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16371378 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384090 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773325 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773287 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773317 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384073 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384097 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773326 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384098 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773337 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16371377 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773292 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773294 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773297 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384096 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773291 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773320 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773340 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773285 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773304 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773303 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773300 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384095 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384086 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773319 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773316 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773311 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384045 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384081 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773296 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773309 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384103 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773322 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773315 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384094 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773318 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16371381 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773306 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16371375 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773299 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array 7934960 - -
Affymetrix Human Gene 1.1 ST Array 7934959 - -
Affymetrix Human Transcriptome Array 2.0 PSR10019065.hg.1 ENST00000362127.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10019065.hg.1 NR_029524.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10019065.hg.1 TC10001508.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1000011383.hg.1 ENST00000362127.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011383.hg.1 NR_029524.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006563.hg.1 ENST00000362154.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006563.hg.1 NR_029519.1 Refseq
Affymetrix Human Clariom D Assay TC1000011383.hg.1 ENST00000362127.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011383.hg.1 NR_029524.1 Refseq
Affymetrix Human Clariom D Assay TC2000006563.hg.1 ENST00000362154.1 Ensembl
Affymetrix Human Clariom D Assay TC2000006563.hg.1 NR_029519.1 Refseq
Affymetrix Human Clariom D Assay PSR1000177364.hg.1 ENST00000362127.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000177364.hg.1 NR_029524.1 Refseq
Affymetrix Human Clariom D Assay PSR1000177364.hg.1 ENST00000362127.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000177364.hg.1 NR_029524.1 Refseq
Affymetrix Human Clariom D Assay PSR1000177364.hg.1 TC1000011383.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3309779 NR_029524.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_3309779 AF480550.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3309779 NR_029524.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3309779 AF480550.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3309779 NR_029524.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3309779 AF480550.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3309779 NR_029524.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3309779 AF480550.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-695c48667f-7flmd:80/100.66.73.211:80.
git-commit: c086736f0d622c41bbf3ea4d71b1c2b8619a5b9d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.35.0-Offline