This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04234032_m1
See other MGST2 GE Assays ›
Gene Symbol:
MGST2
Gene Name:
microsomal glutathione S-transferase 2
Gene Aliases:
GST2, MGST-II
Chromosome Location:
Chr.4: 139665725 - 139754546 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04234032_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MGST2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001204366.1 NP_001191295.1 1 - 2 310 66
NM_001204368.1 NP_001191297.1 1 - 2 310 66
NM_002413.4 NP_002404.1 1 - 2 310 66
XM_017008211.1 XP_016863700.1 1 - 2 322 66
XM_017008212.1 XP_016863701.1 1 - 2 322 66
XM_017008213.1 XP_016863702.1 1 - 2 322 66
GenBank mRNA AA122237.1 - 1 - 2 310 66
AK130948.1 - 1 - 2 259 66
BC025416.1 - 1 - 2 224 3937750 66
BE312976.1 - 1 - 2 274 66
CR407640.1 - 1 - 2 58 66
DB257575.1 - 1 - 2 73 66
U77604.1 - 1 - 2 69 66

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MGST2

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

glutathione biosynthetic process
xenobiotic metabolic process
response to organonitrogen compound
leukotriene biosynthetic process
response to lipopolysaccharide
positive regulation of catalytic activity
oxidation-reduction process
cellular oxidant detoxification
glutathione derivative biosynthetic process
glutathione transferase activity
leukotriene-C4 synthase activity
glutathione peroxidase activity
enzyme activator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline