This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04259568_s1
See other GJB1 GE Assays ›
Gene Symbol:
GJB1
Gene Name:
gap junction protein beta 1
Gene Aliases:
CMTX, CMTX1, CX32
Chromosome Location:
Chr.X: 71215212 - 71225215 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04259568_s1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GJB1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000166.5 NP_000157.1 2 943 151
NM_001097642.2 NP_001091111.1 2 892 151
XM_011530907.2 XP_011529209.1 2 991 151
XM_017029408.1 XP_016884897.1 3 1112 151
GenBank mRNA BC002805.1 - 2 852 3638623 151
BC022426.1 - 2 923 4252281 151
BC039198.1 - 2 865 4710239 151

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00007559.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X000384.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GJB1

Panther Classification:

Molecular Function -

gap junction

Gene Ontology Categories:

Function(s) Process(es)

transport
cell-cell signaling
nervous system development
purine ribonucleotide transport
gap junction assembly
protein oligomerization
transmembrane transport
gap junction channel activity
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000447581.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000447581.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_001097642.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_001097642.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_001097642.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_001097642.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X005107.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X005107.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X005107.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X005107.hg.1 NM_000166.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X005107.hg.1 NM_001097642.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X005107.hg.1 TC0X000384.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0X00151911.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151911.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151911.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151911.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay PSR0X00151911.hg.1 NM_001097642.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00151911.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151911.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151911.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151911.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay PSR0X00151911.hg.1 NM_001097642.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00151911.hg.1 TC0X00007559.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000447581.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000447581.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_001097642.2 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_001097642.2 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_001097642.2 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_001097642.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline