This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04273350_s1
See other MIR651 GE Assays ›
Gene Symbol:
MIR651
Gene Name:
microRNA 651
Gene Aliases:
MIRN651, hsa-mir-651, mir-651
Chromosome Location:
Chr.X: 8126965 - 8127061 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04273350_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR651
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_030380.1 1 48 95

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream patatin like phospholipase domain containing 4 PNPLA4 DXS1283E,
GS2,
iPLA2eta
-198.4kb Anti-sense Chr. X - 7898763 - 7928587 on NCBI Build GRCh38
Downstream variable charge,
X-linked 2
VCX2 VCX-2r,
VCX2R,
VCXB
42.9kb Anti-sense Chr. X - 8169944 - 8171267 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC0X00006563.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X000029.hg.1

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17315631 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276449 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276453 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276457 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276476 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276445 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276472 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572245 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572243 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572242 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572274 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572250 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276468 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276460 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276481 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572275 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572224 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572247 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572248 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572249 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276443 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276473 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572255 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572270 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276441 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276448 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572228 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572257 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276467 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276464 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572231 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572253 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276483 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572244 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572225 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572280 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276439 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276447 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276482 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276466 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276465 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276470 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572255 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572250 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276466 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17315631 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572242 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276473 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572247 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276453 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572257 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572245 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572270 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276465 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276464 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276439 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276460 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276447 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572228 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572243 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276443 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572248 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572244 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276441 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276476 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572253 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572225 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276448 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276467 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572231 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572275 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276470 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276449 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276445 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572280 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572274 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276468 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572224 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276457 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276472 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276481 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11572249 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276483 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32276482 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR0X000366.hg.1 TC0X000029.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X000366.hg.1 ENST00000384896.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X000366.hg.1 NR_030380.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006563.hg.1 ENST00000384896.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006563.hg.1 NR_030380.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006563.hg.1 ENST00000384896.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00006563.hg.1 NR_030380.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00144846.hg.1 ENST00000384896.2 Ensembl
Affymetrix Human Clariom D Assay PSR0X00144846.hg.1 NR_030380.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00144846.hg.1 ENST00000384896.2 Ensembl
Affymetrix Human Clariom D Assay PSR0X00144846.hg.1 NR_030380.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00144846.hg.1 TC0X00006563.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3310504 NR_030380.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3310504 NR_030380.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3310504 NR_030380.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3310504 NR_030380.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-ff4qj:80/100.66.78.114:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0