This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04273469_s1
See other MIR1913 GE Assays ›
Gene Symbol:
MIR1913
Gene Name:
microRNA 1913
Gene Aliases:
MIRN1913, hsa-mir-1913
Chromosome Location:
Chr.6: 166509354 - 166509433 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04273469_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR1913
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_031734.1 1 40 63

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RPS6KA2 intronic transcript 1 RPS6KA2-IT1 -44.0kb Sense Chr. 6 - 166460663 - 166465383 on NCBI Build GRCh38
Downstream RPS6KA2 antisense RNA 1 RPS6KA2-AS1 394.3kb Anti-sense Chr. 6 - 166903698 - 166905069 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC0600013897.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06002310.hg.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242070 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15888529 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242098 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25447287 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25447270 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25447283 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15888520 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15888522 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25447268 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242088 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242100 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242093 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242060 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242083 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25447261 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25447286 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242077 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242086 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15888525 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242090 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25447280 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242059 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242065 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15888512 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242075 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242063 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25447243 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25447276 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242068 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242072 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25447235 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25447237 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242074 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242081 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15888521 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15888520 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242081 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242070 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242077 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242063 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242093 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242090 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242100 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242072 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242060 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25447287 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242059 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15888522 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242083 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15888521 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25447237 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25447276 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25447235 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25447261 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25447270 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242088 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15888512 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25447243 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242086 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242098 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15888529 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242068 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242065 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25447268 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25447286 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25447283 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242074 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05242075 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25447280 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15888525 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600013897.hg.1 ENST00000411026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013897.hg.1 NR_031734.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06029035.hg.1 ENST00000411026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06029035.hg.1 NR_031734.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06029035.hg.1 TC06002310.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0600195825.hg.1 ENST00000411026.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600195825.hg.1 NR_031734.1 Refseq
Affymetrix Human Clariom D Assay PSR0600195825.hg.1 ENST00000411026.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600195825.hg.1 NR_031734.1 Refseq
Affymetrix Human Clariom D Assay PSR0600195825.hg.1 TC0600013897.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0600013897.hg.1 ENST00000411026.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013897.hg.1 NR_031734.1 Refseq
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3308025 NR_031734.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3308025 NR_031734.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308025 NR_031734.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308025 NR_031734.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline