This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04274084_m1
See other USP2-AS1 GE Assays ›
Gene Symbol:
USP2-AS1
Gene Name:
USP2 antisense RNA 1 (head to head)
Gene Aliases:
THY1-AS1
Chromosome Location:
Chr.11: 119381778 - 119499233 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04274084_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: USP2-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_034160.1 3 - 4 472 92
GenBank mRNA AL559224.3 - 3 - 4 424 92
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BG186533.1 - 1 - 2 174 Chr. 11 - 119469828 - 119499162 [+] on Build GRCh38 92

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream Thy-1 cell surface antigen THY1 CD90,
CDw90
-46.3kb Anti-sense Chr. 11 - 119417941 - 119424985 on NCBI Build GRCh38
Downstream nectin cell adhesion molecule 1 NECTIN1 CD111,
CLPED1,
ED4,
HIgR,
HV1S,
HVEC,
OFC7,
PRR,
PRR1,
PVRL1,
PVRR,
PVRR1,
SK-12,
nectin-1
138.9kb Anti-sense Chr. 11 - 119638098 - 119728725 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100013105.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11001092.hg.1

Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Human 1B Microarray G4111A A_32_P849507 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000530002.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000530002.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000530002.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000530002.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000530002.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000578923.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000578923.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 ENST00000578923.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 NR_034160.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 NR_034160.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 NR_034160.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 NR_034160.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 NR_034160.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013105.hg.1 NR_034160.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11013887.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11013887.hg.1 NR_034160.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11013887.hg.1 TC11001092.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11013885.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11013885.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11013885.hg.1 NR_034160.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11013885.hg.1 TC11001092.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11013886.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11013886.hg.1 NR_034160.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11013886.hg.1 TC11001092.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11013883.hg.1 NR_034160.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11013883.hg.1 TC11001092.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11013884.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11013884.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11013884.hg.1 NR_034160.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11013884.hg.1 TC11001092.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1100202790.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202790.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202790.hg.1 NR_034160.1 Refseq
Affymetrix Human Clariom D Assay PSR1100202790.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202790.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202790.hg.1 NR_034160.1 Refseq
Affymetrix Human Clariom D Assay PSR1100202790.hg.1 TC1100013105.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100202795.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100202795.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100202795.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202795.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202795.hg.1 ENST00000530002.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202795.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202795.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202795.hg.1 ENST00000578923.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202795.hg.1 NR_034160.1 Refseq
Affymetrix Human Clariom D Assay PSR1100202795.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100202795.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100202795.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202795.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202795.hg.1 ENST00000530002.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202795.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202795.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202795.hg.1 ENST00000578923.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202795.hg.1 NR_034160.1 Refseq
Affymetrix Human Clariom D Assay PSR1100202795.hg.1 TC1100013105.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100202801.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100202801.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100202801.hg.1 ENST00000530002.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202801.hg.1 NR_034160.1 Refseq
Affymetrix Human Clariom D Assay PSR1100202801.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100202801.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100202801.hg.1 ENST00000530002.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202801.hg.1 NR_034160.1 Refseq
Affymetrix Human Clariom D Assay PSR1100202801.hg.1 TC1100013105.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100202803.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100202803.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100202803.hg.1 ENST00000578923.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202803.hg.1 NR_034160.1 Refseq
Affymetrix Human Clariom D Assay PSR1100202803.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100202803.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100202803.hg.1 ENST00000578923.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202803.hg.1 NR_034160.1 Refseq
Affymetrix Human Clariom D Assay PSR1100202803.hg.1 TC1100013105.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100202796.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100202796.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100202796.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202796.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202796.hg.1 ENST00000530002.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202796.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202796.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202796.hg.1 NR_034160.1 Refseq
Affymetrix Human Clariom D Assay PSR1100202796.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100202796.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100202796.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202796.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202796.hg.1 ENST00000530002.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202796.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202796.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202796.hg.1 NR_034160.1 Refseq
Affymetrix Human Clariom D Assay PSR1100202796.hg.1 TC1100013105.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000530002.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000530002.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000530002.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000530002.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000530002.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000578923.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000578923.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 ENST00000578923.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013105.hg.1 NR_034160.1 Refseq
Affymetrix Human Clariom D Assay TC1100013105.hg.1 NR_034160.1 Refseq
Affymetrix Human Clariom D Assay TC1100013105.hg.1 NR_034160.1 Refseq
Affymetrix Human Clariom D Assay TC1100013105.hg.1 NR_034160.1 Refseq
Affymetrix Human Clariom D Assay TC1100013105.hg.1 NR_034160.1 Refseq
Affymetrix Human Clariom D Assay TC1100013105.hg.1 NR_034160.1 Refseq
Affymetrix Human Clariom D Assay PSR1100202797.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100202797.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100202797.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202797.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202797.hg.1 ENST00000530002.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202797.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202797.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202797.hg.1 ENST00000578923.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202797.hg.1 NR_034160.1 Refseq
Affymetrix Human Clariom D Assay PSR1100202797.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100202797.hg.1 ENST00000498979.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100202797.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202797.hg.1 ENST00000500970.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202797.hg.1 ENST00000530002.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202797.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202797.hg.1 ENST00000577297.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202797.hg.1 ENST00000578923.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100202797.hg.1 NR_034160.1 Refseq
Affymetrix Human Clariom D Assay PSR1100202797.hg.1 TC1100013105.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0