This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04274678_s1
See other MIR4738 GE Assays ›
Gene Symbol:
MIR4738
Gene Name:
microRNA 4738
Gene Aliases:
-
Chromosome Location:
Chr.17: 75784521 - 75784607 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04274678_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR4738
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_039892.1 1 33 78
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BI828778.1 - 1 134 Chr. 17 - 75783856 - 75784709 [-] on Build GRCh38 78
GenBankmRNA AK096931.1 - 1 1318 Chr. 17 - 75783107 - 75785893 [-] on Build GRCh38 78

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream H3 histone,
family 3B
H3F3B H3.3B -4.6kb Sense Chr. 17 - 75776434 - 75779935 on NCBI Build GRCh38
Downstream unkempt family zinc finger UNK UNKEMPT,
ZC3H5,
ZC3HDC5
232.0 Anti-sense Chr. 17 - 75784771 - 75825805 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC1700012471.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17001883.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700012471.hg.1 ENST00000579134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012471.hg.1 ENST00000591893.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012471.hg.1 ENST00000591893.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012471.hg.1 ENST00000591893.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012471.hg.1 NR_039892.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17025314.hg.1 NR_039892.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17025314.hg.1 TC17001883.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1700207925.hg.1 ENST00000579134.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700207925.hg.1 ENST00000591893.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700207925.hg.1 NR_039892.1 Refseq
Affymetrix Human Clariom D Assay PSR1700207925.hg.1 TC1700012471.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1700207925.hg.1 ENST00000579134.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700207925.hg.1 ENST00000591893.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700207925.hg.1 NR_039892.1 Refseq
Affymetrix Human Clariom D Assay TC1700012471.hg.1 ENST00000579134.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012471.hg.1 ENST00000591893.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012471.hg.1 ENST00000591893.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012471.hg.1 ENST00000591893.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012471.hg.1 NR_039892.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline