This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04274954_m1
See other PVRL3-AS1 GE Assays ›
Gene Symbol:
PVRL3-AS1
Gene Name:
PVRL3 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 111045315 - 111069959 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04274954_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PVRL3-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045114.1 2 - 3 385 162
GenBank mRNA BU852869.1 - 388 162
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AA625639.1 - 2 - 3 Chr. 3 - 111046900 - 111069390 [+] on Build GRCh38 162

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream putative uncharacterized protein LOC151760 LOC151760 -81.6kb Sense Chr. 3 - 110892123 - 110974348 on NCBI Build GRCh38
Downstream nectin cell adhesion molecule 3 NECTIN3 CD113,
CDW113,
NECTIN-3,
PPR3,
PRR3,
PVRL3,
PVRR3
2.3kb Anti-sense Chr. 3 - 111071759 - 111201444 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300011974.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001642.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03029625.hg.1 TC03001642.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03029625.hg.1 ENST00000463025.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03029625.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03029625.hg.1 NR_045114.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000463025.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000463025.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000463025.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000474769.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000474769.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000474769.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000474769.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000474769.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000474769.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000474769.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000474769.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000474769.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000474769.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000476301.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000476301.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000476301.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000476301.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000476301.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000476301.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000476301.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000476301.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000476301.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000476301.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 NR_045114.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 NR_045114.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 NR_045114.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 NR_045114.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 NR_045114.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 NR_045114.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011974.hg.1 NR_045114.1 Refseq
Affymetrix Human Clariom D Assay PSR0300189121.hg.1 ENST00000463025.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300189121.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300189121.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300189121.hg.1 NR_045114.1 Refseq
Affymetrix Human Clariom D Assay PSR0300189121.hg.1 ENST00000463025.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300189121.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300189121.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300189121.hg.1 NR_045114.1 Refseq
Affymetrix Human Clariom D Assay PSR0300189121.hg.1 TC0300011974.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000463025.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000463025.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000463025.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000467426.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000474769.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000474769.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000474769.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000474769.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000474769.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000474769.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000474769.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000474769.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000474769.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000474769.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000476301.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000476301.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000476301.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000476301.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000476301.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000476301.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000476301.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000476301.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000476301.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000476301.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 ENST00000491709.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011974.hg.1 NR_045114.1 Refseq
Affymetrix Human Clariom D Assay TC0300011974.hg.1 NR_045114.1 Refseq
Affymetrix Human Clariom D Assay TC0300011974.hg.1 NR_045114.1 Refseq
Affymetrix Human Clariom D Assay TC0300011974.hg.1 NR_045114.1 Refseq
Affymetrix Human Clariom D Assay TC0300011974.hg.1 NR_045114.1 Refseq
Affymetrix Human Clariom D Assay TC0300011974.hg.1 NR_045114.1 Refseq
Affymetrix Human Clariom D Assay TC0300011974.hg.1 NR_045114.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0