This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04276267_m1
See other RIC3 GE Assays ›
Gene Symbol:
RIC3
Gene Name:
RIC3 acetylcholine receptor chaperone
Gene Aliases:
AYST720, PRO1385
Chromosome Location:
Chr.11: 8092965 - 8169088 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04276267_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RIC3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001206671.2 NP_001193600.1 1 - 2 177 104
NM_001206672.2 NP_001193601.1 1 - 2 177 104
NM_024557.4 NP_078833.3 1 - 2 177 104
XM_006718317.3 XP_006718380.1 1 - 2 222 104
XM_006718318.3 XP_006718381.1 1 - 2 222 104
XM_006718321.3 XP_006718384.1 1 - 2 222 104
XM_017018287.1 XP_016873776.1 1 - 2 222 104
XM_017018290.1 XP_016873779.1 1 - 2 222 104
GenBank mRNA AK293933.1 - 1 - 2 140 104
AK315379.1 - 1 - 2 136 104
AM422214.1 - 1 - 2 123 104
AY326435.1 - 1 - 2 123 104
AY326436.1 - 1 - 2 158 104
AY358475.1 - 1 - 2 148 104
DC420812.1 - 1 - 2 177 104
KU178773.1 - 1 - 2 123 104
KU178774.1 - 1 - 2 123 104

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RIC3

Gene Ontology Categories:

Function(s) Process(es)

protein folding
positive regulation of cytosolic calcium ion concentration
synaptic transmission, cholinergic
protein localization to cell surface
cellular protein complex assembly
acetylcholine receptor binding
protein binding involved in protein folding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0