This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04329622_m1
See other IKZF3 GE Assays ›
Gene Symbol:
IKZF3
Gene Name:
IKAROS family zinc finger 3
Gene Aliases:
AIO, AIOLOS, ZNFN1A3
Chromosome Location:
Chr.17: 39757715 - 39864188 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04329622_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IKZF3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001257414.1 NP_001244343.1 3 - 4 219 76
NM_001284514.1 NP_001271443.1 3 - 4 219 76
NM_001284515.1 NP_001271444.1 3 - 4 219 76
NM_012481.4 NP_036613.2 3 - 4 219 76
NM_183228.2 NP_899051.1 3 - 4 219 76
NM_183229.2 NP_899052.1 3 - 4 219 76
NM_183230.2 NP_899053.1 3 - 4 219 76
NM_183231.2 NP_899054.1 3 - 4 219 76
NM_183232.2 NP_899055.1 3 - 4 219 76
GenBank mRNA AF129512.1 - 3 - 4 219 76
AJ292565.1 - 2 - 3 157 76
AJ292566.1 - 2 - 3 157 76
AJ292567.1 - 2 - 3 157 76
AJ292568.1 - 2 - 3 157 76
AJ292569.1 - 2 - 3 157 76
AY377980.1 - 3 - 4 219 76
AY377981.1 - 3 - 4 219 76
AY377982.1 - 3 - 4 219 76
BC032707.1 - 3 - 4 208 5497068 76

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001284514.1, NM_001284515.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IKZF3

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
mesoderm development
regulation of B cell proliferation
B cell activation
regulation of apoptotic process
regulation of B cell differentiation
regulation of lymphocyte differentiation
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein homodimerization activity
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0