| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08018423.hg.1 |
ENST00000504861.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08018423.hg.1 |
NR_046114.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08018423.hg.1 |
TC08001420.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012442.hg.1 |
ENST00000521148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012442.hg.1 |
ENST00000521148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012442.hg.1 |
ENST00000521148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012442.hg.1 |
ENST00000521148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012442.hg.1 |
ENST00000521148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012442.hg.1 |
ENST00000521148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012442.hg.1 |
ENST00000521148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012442.hg.1 |
ENST00000521148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012442.hg.1 |
ENST00000521230.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012442.hg.1 |
ENST00000521230.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012442.hg.1 |
ENST00000521230.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012442.hg.1 |
ENST00000521230.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012442.hg.1 |
ENST00000521230.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012442.hg.1 |
ENST00000521230.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012442.hg.1 |
ENST00000521230.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012442.hg.1 |
ENST00000521230.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012442.hg.1 |
NR_046114.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012442.hg.1 |
NR_046114.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012442.hg.1 |
NR_046114.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012442.hg.1 |
NR_046114.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012442.hg.1 |
NR_046114.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08018420.hg.1 |
ENST00000504861.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08018420.hg.1 |
ENST00000521148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08018420.hg.1 |
ENST00000521230.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08018420.hg.1 |
NR_046114.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08018420.hg.1 |
TC08001420.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0800187681.hg.1 |
NR_046114.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800187681.hg.1 |
NR_046114.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800187681.hg.1 |
TC0800012442.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800012442.hg.1 |
ENST00000521148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012442.hg.1 |
ENST00000521148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012442.hg.1 |
ENST00000521148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012442.hg.1 |
ENST00000521148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012442.hg.1 |
ENST00000521148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012442.hg.1 |
ENST00000521148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012442.hg.1 |
ENST00000521148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012442.hg.1 |
ENST00000521148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012442.hg.1 |
ENST00000521230.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012442.hg.1 |
ENST00000521230.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012442.hg.1 |
ENST00000521230.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012442.hg.1 |
ENST00000521230.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012442.hg.1 |
ENST00000521230.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012442.hg.1 |
ENST00000521230.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012442.hg.1 |
ENST00000521230.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012442.hg.1 |
ENST00000521230.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012442.hg.1 |
NR_046114.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012442.hg.1 |
NR_046114.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012442.hg.1 |
NR_046114.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012442.hg.1 |
NR_046114.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012442.hg.1 |
NR_046114.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800187682.hg.1 |
NR_046114.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800187682.hg.1 |
NR_046114.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800187682.hg.1 |
TC0800012442.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800187674.hg.1 |
ENST00000521148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800187674.hg.1 |
ENST00000521148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800187674.hg.1 |
ENST00000521230.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800187674.hg.1 |
ENST00000521230.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800187674.hg.1 |
NR_046114.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800187674.hg.1 |
ENST00000521148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800187674.hg.1 |
ENST00000521148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800187674.hg.1 |
ENST00000521230.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800187674.hg.1 |
ENST00000521230.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800187674.hg.1 |
NR_046114.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800187674.hg.1 |
TC0800012442.hg.1 |
Clariom D TC |