This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04332205_m1
See other RBPMS-AS1 GE Assays ›
Gene Symbol:
RBPMS-AS1
Gene Name:
RBPMS antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.8: 30382119 - 30385401 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04332205_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBPMS-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_046205.1 2 - 3 412 100
GenBank mRNA AK057935.1 - 2 - 3 412 102

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream dynactin subunit 6 DCTN6 WS-3,
WS3,
p27
-198.5kb Anti-sense Chr. 8 - 30156297 - 30183639 on NCBI Build GRCh38
Downstream RNA binding protein with multiple splicing RBPMS HERMES 768.0 Anti-sense Chr. 8 - 30384501 - 30572261 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800010030.hg.1

Set Membership:

Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 212082 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P35506 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P58565 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 11732 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 24078 - -
Agilent Human Genome CGH + SNP Microarray A_16_P01889379 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3316903 - -
Whole Human Genome Oligo Microarray G4112A A_32_P35506 - -
Whole Human Genome Oligo Microarray G4112A A_24_P58565 - -
Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P35506 - -
Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P58565 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01889379 - -
Agilent Human 1B Microarray G4111A A_32_P35504 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800010030.hg.1 ENST00000517521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010030.hg.1 ENST00000517521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010030.hg.1 ENST00000517521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010030.hg.1 ENST00000517521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010030.hg.1 ENST00000517521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010030.hg.1 ENST00000517521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010030.hg.1 ENST00000517521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010030.hg.1 ENST00000517521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010030.hg.1 ENST00000519753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010030.hg.1 ENST00000519753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010030.hg.1 ENST00000519753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010030.hg.1 ENST00000519753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010030.hg.1 ENST00000523643.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010030.hg.1 ENST00000523643.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010030.hg.1 ENST00000523643.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010030.hg.1 ENST00000523643.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010030.hg.1 ENST00000523643.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010030.hg.1 ENST00000523643.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010030.hg.1 NR_046205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010030.hg.1 NR_046205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010030.hg.1 NR_046205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010030.hg.1 NR_046205.1 Refseq
Affymetrix Human Clariom D Assay TC0800010030.hg.1 ENST00000517521.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010030.hg.1 ENST00000517521.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010030.hg.1 ENST00000517521.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010030.hg.1 ENST00000517521.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010030.hg.1 ENST00000517521.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010030.hg.1 ENST00000517521.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010030.hg.1 ENST00000517521.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010030.hg.1 ENST00000517521.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010030.hg.1 ENST00000519753.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010030.hg.1 ENST00000519753.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010030.hg.1 ENST00000519753.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010030.hg.1 ENST00000519753.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010030.hg.1 ENST00000523643.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010030.hg.1 ENST00000523643.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010030.hg.1 ENST00000523643.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010030.hg.1 ENST00000523643.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010030.hg.1 ENST00000523643.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010030.hg.1 ENST00000523643.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010030.hg.1 NR_046205.1 Refseq
Affymetrix Human Clariom D Assay TC0800010030.hg.1 NR_046205.1 Refseq
Affymetrix Human Clariom D Assay TC0800010030.hg.1 NR_046205.1 Refseq
Affymetrix Human Clariom D Assay TC0800010030.hg.1 NR_046205.1 Refseq
Affymetrix Human Clariom D Assay PSR0800168267.hg.1 ENST00000517521.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800168267.hg.1 ENST00000517521.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800168267.hg.1 ENST00000519753.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800168267.hg.1 ENST00000523643.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800168267.hg.1 ENST00000523643.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800168267.hg.1 NR_046205.1 Refseq
Affymetrix Human Clariom D Assay PSR0800168267.hg.1 ENST00000517521.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800168267.hg.1 ENST00000517521.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800168267.hg.1 ENST00000519753.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800168267.hg.1 ENST00000523643.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800168267.hg.1 ENST00000523643.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800168267.hg.1 NR_046205.1 Refseq
Affymetrix Human Clariom D Assay PSR0800168267.hg.1 TC0800010030.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3190337 AK057935.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_3266802 AK057935.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline