This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04332438_g1
See other AOAH-IT1 GE Assays ›
Gene Symbol:
AOAH-IT1
Gene Name:
AOAH intronic transcript 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 36597834 - 36600120 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04332438_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AOAH-IT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_046764.1 2 - 3 528 77

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream anillin actin binding protein ANLN FSGS8,
Scraps,
scra
-144.0kb Anti-sense Chr. 7 - 36389803 - 36453791 on NCBI Build GRCh38
Downstream engulfment and cell motility 1 ELMO1 CED-12,
CED12,
ELMO-1
254.6kb Sense Chr. 7 - 36852906 - 37449409 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700010754.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700010754.hg.1 ENST00000449672.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010754.hg.1 ENST00000449672.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010754.hg.1 ENST00000449672.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010754.hg.1 NR_046764.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010754.hg.1 NR_046764.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010754.hg.1 NR_046764.1 Refseq
Affymetrix Human Clariom D Assay TC0700010754.hg.1 ENST00000449672.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010754.hg.1 ENST00000449672.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010754.hg.1 ENST00000449672.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010754.hg.1 NR_046764.1 Refseq
Affymetrix Human Clariom D Assay TC0700010754.hg.1 NR_046764.1 Refseq
Affymetrix Human Clariom D Assay TC0700010754.hg.1 NR_046764.1 Refseq
Affymetrix Human Clariom D Assay PSR0700176585.hg.1 ENST00000449672.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700176585.hg.1 NR_046764.1 Refseq
Affymetrix Human Clariom D Assay PSR0700176585.hg.1 ENST00000449672.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700176585.hg.1 NR_046764.1 Refseq
Affymetrix Human Clariom D Assay PSR0700176585.hg.1 TC0700010754.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline