| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013893.hg.1 |
ENST00000416770.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013893.hg.1 |
ENST00000416770.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013893.hg.1 |
ENST00000416770.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013893.hg.1 |
ENST00000416770.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013893.hg.1 |
NR_046793.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013893.hg.1 |
NR_046793.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013893.hg.1 |
NR_046793.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013893.hg.1 |
NR_046793.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600195820.hg.1 |
ENST00000416770.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600195820.hg.1 |
NR_046793.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600195820.hg.1 |
ENST00000416770.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600195820.hg.1 |
NR_046793.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600195820.hg.1 |
TC0600013893.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600013893.hg.1 |
ENST00000416770.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013893.hg.1 |
ENST00000416770.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013893.hg.1 |
ENST00000416770.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013893.hg.1 |
ENST00000416770.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013893.hg.1 |
NR_046793.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013893.hg.1 |
NR_046793.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013893.hg.1 |
NR_046793.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013893.hg.1 |
NR_046793.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600195821.hg.1 |
ENST00000416770.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600195821.hg.1 |
NR_046793.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600195821.hg.1 |
ENST00000416770.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600195821.hg.1 |
NR_046793.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600195821.hg.1 |
TC0600013893.hg.1 |
Clariom D TC |