This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04332444_m1
See other RPS6KA2-IT1 GE Assays ›
Gene Symbol:
RPS6KA2-IT1
Gene Name:
RPS6KA2 intronic transcript 1
Gene Aliases:
-
Chromosome Location:
Chr.6: 166460663 - 166465383 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04332444_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPS6KA2-IT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_046793.1 3 - 4 405 102
GenBank mRNA AW992696.1 - 3 - 4 411 102

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream mitochondrial pyruvate carrier 1 MPC1 BRP44L,
CGI-129,
MPYCD,
dJ68L15.3
-77.7kb Sense Chr. 6 - 166364919 - 166383013 on NCBI Build GRCh38
Downstream microRNA 1913 MIR1913 MIRN1913,
hsa-mir-1913
46.2kb Sense Chr. 6 - 166509354 - 166509433 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600013893.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600013893.hg.1 ENST00000416770.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013893.hg.1 ENST00000416770.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013893.hg.1 ENST00000416770.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013893.hg.1 ENST00000416770.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013893.hg.1 NR_046793.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013893.hg.1 NR_046793.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013893.hg.1 NR_046793.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013893.hg.1 NR_046793.1 Refseq
Affymetrix Human Clariom D Assay PSR0600195818.hg.1 ENST00000416770.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600195818.hg.1 NR_046793.1 Refseq
Affymetrix Human Clariom D Assay PSR0600195818.hg.1 ENST00000416770.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600195818.hg.1 NR_046793.1 Refseq
Affymetrix Human Clariom D Assay PSR0600195818.hg.1 TC0600013893.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0600013893.hg.1 ENST00000416770.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013893.hg.1 ENST00000416770.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013893.hg.1 ENST00000416770.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013893.hg.1 ENST00000416770.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013893.hg.1 NR_046793.1 Refseq
Affymetrix Human Clariom D Assay TC0600013893.hg.1 NR_046793.1 Refseq
Affymetrix Human Clariom D Assay TC0600013893.hg.1 NR_046793.1 Refseq
Affymetrix Human Clariom D Assay TC0600013893.hg.1 NR_046793.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline