Assay Details
| Gene Symbol: MIR5704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049890.1 | 1 | 38 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | BCL2 like 12 pseudogene 1 | BCL2L12P1 | -458.7kb | Anti-sense | Chr. 3 - 131526468 - 131527141 on NCBI Build GRCh38 | |
| Downstream |
acid phosphatase, prostate |
ACPP |
5'-NT, ACP-3, ACP3 |
331.4kb | Anti-sense | Chr. 3 - 132317367 - 132368765 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0300012454.hg.1
Set Membership: |
|

MIR5704
Genomic Map