This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04332732_m1
See other NRM GE Assays ›
Gene Symbol:
NRM
Gene Name:
nurim (nuclear envelope membrane protein)
Gene Aliases:
NRM29
Chromosome Location:
Chr.6: 30688047 - 30691420 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04332732_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NRM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_073066.1 1 - 2 290 69
GenBank mRNA AK298152.1 - 1 - 2 147 69
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX375666.2 - 1 - 2 128 Chr. 6 - 30688304 - 30690967 [-] on Build GRCh38 69
GenBankmRNA DC360142.1 - 1 - 2 Chr. 6 - 30688749 - 30690983 [-] on Build GRCh38 70
GenBankmRNA BG256226.1 - 107 Chr. - - [-] on Build 69

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream protein phosphatase 1 regulatory subunit 18 PPP1R18 HKMT1098,
KIAA1949
-1.4kb Sense Chr. 6 - 30676389 - 30687895 on NCBI Build GRCh38
Downstream mediator of DNA damage checkpoint 1 MDC1 NFBD1 8.7kb Sense Chr. 6 - 30699807 - 30717988 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK298152.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0