This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04333964_g1
See other OBSCN GE Assays ›
Gene Symbol:
OBSCN
Gene Name:
obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
Gene Aliases:
ARHGEF30, UNC89
Chromosome Location:
Chr.1: 228208013 - 228381431 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04333964_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OBSCN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001098623.2 NP_001092093.2 39 - 40 10680 77
NM_001271223.2 NP_001258152.2 44 - 45 11967 77
NM_052843.3 NP_443075.3 39 - 40 10680 77
XM_005273287.4 XP_005273344.1 43 - 44 11771 77
XM_005273291.4 XP_005273348.1 43 - 44 11771 77
XM_005273298.4 XP_005273355.1 43 - 44 11771 77
XM_005273307.4 XP_005273364.1 43 - 44 11771 77
XM_006711822.3 XP_006711885.1 43 - 44 11771 77
XM_006711823.3 XP_006711886.1 43 - 44 11771 77
XM_006711827.3 XP_006711890.1 43 - 44 11770 77
XM_006711829.3 XP_006711892.1 42 - 43 11494 77
XM_011544297.2 XP_011542599.1 43 - 44 11771 77
XM_011544299.2 XP_011542601.1 42 - 43 11495 77
XM_017002443.1 XP_016857932.1 43 - 44 11771 77
XM_017002444.1 XP_016857933.1 43 - 44 11734 77
XM_017002445.1 XP_016857934.1 43 - 44 11771 77
XM_017002446.1 XP_016857935.1 43 - 44 11771 77
XM_017002447.1 XP_016857936.1 43 - 44 11771 77
XM_017002448.1 XP_016857937.1 43 - 44 11621 77
XM_017002449.1 XP_016857938.1 43 - 44 11772 77
XM_017002450.1 XP_016857939.1 43 - 44 11772 77
XM_017002452.1 XP_016857941.1 43 - 44 11772 77
XM_017002453.1 XP_016857942.1 42 - 43 11495 77
XM_017002454.1 XP_016857943.1 42 - 43 11495 77
XM_017002455.1 XP_016857944.1 42 - 43 11495 77
XM_017002456.1 XP_016857945.1 42 - 43 11495 77
XM_017002457.1 XP_016857946.1 42 - 43 11495 77
XM_017002458.1 XP_016857947.1 43 - 44 11772 77
XM_017002459.1 XP_016857948.1 43 - 44 11772 77
XM_017002461.1 XP_016857950.1 43 - 44 11771 77
XM_017002462.1 XP_016857951.1 41 - 42 11219 77
XM_017002463.1 XP_016857952.1 41 - 42 11198 77
XM_017002464.1 XP_016857953.1 43 - 44 11771 77
XM_017002465.1 XP_016857954.1 43 - 44 11772 77
XM_017002466.1 XP_016857955.1 43 - 44 11772 77
XM_017002467.1 XP_016857956.1 43 - 44 11772 77
XM_017002468.1 XP_016857957.1 43 - 44 11772 77
XM_017002469.1 XP_016857958.1 43 - 44 11772 77
XM_017002471.1 XP_016857960.1 43 - 44 11770 77
XM_017002472.1 XP_016857961.1 43 - 44 11770 77
XM_017002473.1 XP_016857962.1 43 - 44 11770 77
GenBank mRNA AB046776.1 - 9 - 10 2433 77
AJ002535.1 - 40 - 41 10677 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100011861.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01001888.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene OBSCN

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
multicellular organism development
regulation of Rho protein signal transduction
protein localization to M-band
positive regulation of apoptotic process
positive regulation of GTPase activity
sarcomere organization
regulation of small GTPase mediated signal transduction
mitophagy in response to mitochondrial depolarization
protein serine/threonine kinase activity
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
protein binding
calmodulin binding
ATP binding
structural constituent of muscle
ankyrin binding
titin binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01029420.hg.1 TC01001888.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01029420.hg.1 ENST00000284548.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01029420.hg.1 ENST00000359599.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01029420.hg.1 ENST00000366707.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01029420.hg.1 ENST00000366709.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01029420.hg.1 ENST00000422127.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01029420.hg.1 ENST00000483539.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01029420.hg.1 NM_001098623.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01029420.hg.1 NM_052843.3 Refseq
Affymetrix Human Clariom D Assay PSR0100187575.hg.1 ENST00000284548.15 Ensembl
Affymetrix Human Clariom D Assay PSR0100187575.hg.1 ENST00000366707.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100187575.hg.1 ENST00000422127.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100187575.hg.1 ENST00000483539.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100187575.hg.1 ENST00000570156.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100187575.hg.1 NM_001098623.2 Refseq
Affymetrix Human Clariom D Assay PSR0100187575.hg.1 NM_001271223.2 Refseq
Affymetrix Human Clariom D Assay PSR0100187575.hg.1 NM_052843.3 Refseq
Affymetrix Human Clariom D Assay PSR0100187575.hg.1 ENST00000284548.15 Ensembl
Affymetrix Human Clariom D Assay PSR0100187575.hg.1 ENST00000366707.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100187575.hg.1 ENST00000422127.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100187575.hg.1 ENST00000483539.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100187575.hg.1 ENST00000570156.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100187575.hg.1 NM_001098623.2 Refseq
Affymetrix Human Clariom D Assay PSR0100187575.hg.1 NM_001271223.2 Refseq
Affymetrix Human Clariom D Assay PSR0100187575.hg.1 NM_052843.3 Refseq
Affymetrix Human Clariom D Assay PSR0100187575.hg.1 TC0100011861.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0